Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012707.1 | - | 1 - 2 | 279 | 106 | |
| BC021396.1 | - | 1 - 2 | 238 | 106 | ||
| U96963.1 | - | 1 - 2 | 211 | 106 | ||
More Information
Set Membership: |
|

Diap1
Genomic Map