Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK156391.1 | - | 9 - 10 | 1309 | 82 | |
| AK220466.1 | - | 22 - 23 | 3058 | 82 | ||
| BC070412.1 | - | 23 - 24 | 3223 | 5705112 | 82 | |
| U96963.1 | - | 21 - 22 | 3061 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap1
Genomic Map