Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK135915.1 | - | 1 - 2 | 147 | 114 | |
| AK156391.1 | - | 12 - 13 | 1731 | 114 | ||
| AK220466.1 | - | 25 - 26 | 3480 | 114 | ||
| BC070412.1 | - | 26 - 27 | 3645 | 5705112 | 114 | |
| U96963.1 | - | 24 - 25 | 3483 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap1
Genomic Map