Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140734.1 | - | 4 - 5 | 457 | 73 | |
| AK220466.1 | - | 4 - 5 | 466 | 73 | ||
| BC021396.1 | - | 3 - 4 | 496 | 73 | ||
| BC070412.1 | - | 5 - 6 | 631 | 5705112 | 73 | |
| U96963.1 | - | 3 - 4 | 469 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap1
Genomic Map