Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140734.1 | - | 7 - 8 | 739 | 72 | |
| AK220466.1 | - | 7 - 8 | 748 | 72 | ||
| BC021396.1 | - | 6 - 7 | 778 | 72 | ||
| BC070412.1 | - | 8 - 9 | 913 | 5705112 | 72 | |
| U96963.1 | - | 6 - 7 | 751 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap1
Genomic Map