Assay Details
| Gene Symbol: Slc26a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011867.3 | NP_035997.1 | 11 - 12 | 1550 | 104 | |
| GenBank mRNA | AF167411.1 | - | 11 - 12 | 1551 | 104 | |
| BC140995.1 | - | 10 - 11 | 1398 | 9055607 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a4
Genomic Map