Assay Details
| Gene Symbol: Ndfip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050560.1 | - | 2 - 3 | 292 | 76 | |
| AK179021.1 | - | 1 - 2 | 96 | 76 | ||
| AK194676.1 | - | 1 - 2 | 96 | 76 | ||
| AK196952.1 | - | 1 - 2 | 96 | 76 | ||
| AK200634.1 | - | 198 | 76 | |||
More Information
Set Membership: |
|

Ndfip1
Genomic Map