Assay Details
| Gene Symbol: Calr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033226.4 | NP_001028398.1 | 2 - 3 | 450 | 65 | |
| NM_001285895.1 | NP_001272824.1 | 3 - 4 | 414 | 65 | ||
| NM_001285896.1 | NP_001272825.1 | 3 - 4 | 427 | 65 | ||
| NM_001285897.1 | NP_001272826.1 | 3 - 4 | 669 | 65 | ||
| NM_001285898.1 | NP_001272827.1 | 3 - 4 | 679 | 65 | ||
| GenBank mRNA | AK077094.1 | - | 3 - 4 | 519 | 65 | |
| AK139540.1 | - | 3 - 4 | 429 | 65 | ||
| AK163350.1 | - | 2 - 3 | 451 | 65 | ||
| BC044740.2 | - | 3 - 4 | 679 | 65 | ||
| JN255799.1 | - | 3 - 4 | 414 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001285896.1, NM_001285897.1, NM_001285898.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077593 | BC044740.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077593 | AK077094.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077593 | AK139540.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 960592 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720320 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 388281 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905209 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829881 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905782 | AK004527.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905782 | AK009179.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905782 | AK154343.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905782 | AK158979.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905782 | AK204928.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905782 | BC036386.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905782 | NM_026452.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529864 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72937 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 463004 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482671 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 806929 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 149207 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53403 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99804 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077593 | BC044740.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077593 | AK077094.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077593 | AK139540.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498475 | AK163350.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498475 | BC044740.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498475 | AK077094.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498475 | AK139540.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498475 | NM_001033226.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23200011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23200019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23200023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23199987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23199993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23200013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23200021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23200017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23200004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23200010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23199995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23200001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23199999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23199992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23200008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23200026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23199991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23200009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23199989 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00957839 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05355805 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00957799 | - | - |
| Whole Mouse Genome G4122A | A_51_P498475 | AK163350.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P498475 | BC044740.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P498475 | AK077094.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P498475 | AK139540.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P498475 | NM_001033226.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00957839 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00957799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23200103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23199976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199989 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04078878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03592567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03429125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03630672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03450685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03633083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03628979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03902389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03642832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03322088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03644658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03306494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04078870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03429129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03628980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03572524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03435498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03902388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23199978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04024018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03661957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23200115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03215300 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10506935 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506936 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506937 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506938 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506932 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506933 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506931 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10506929 | - | - |
| Mouse Genome 430 2.0 Array | 1419843_at | - | - |
| Mouse Genome 430 2.0 Array | 1419843_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400008377.mm.1 | NM_001033226.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008377.mm.1 | NM_001285895.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008377.mm.1 | NM_001285896.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008377.mm.1 | NM_001285897.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008377.mm.1 | NM_001285898.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008377.mm.1 | TC0400001163.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 630995 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102690484 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226822 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226822 | - | - |

Calr4
Genomic Map