Assay Details
| Gene Symbol: Calr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033226.4 | NP_001028398.1 | 3 - 4 | 542 | 97 | |
| NM_001285895.1 | NP_001272824.1 | 4 - 5 | 506 | 97 | ||
| NM_001285896.1 | NP_001272825.1 | 4 - 5 | 519 | 97 | ||
| NM_001285897.1 | NP_001272826.1 | 4 - 5 | 761 | 97 | ||
| NM_001285898.1 | NP_001272827.1 | 4 - 5 | 771 | 97 | ||
| GenBank mRNA | AK077094.1 | - | 4 - 5 | 611 | 97 | |
| AK139540.1 | - | 4 - 5 | 521 | 97 | ||
| AK163350.1 | - | 3 - 4 | 543 | 97 | ||
| BC044740.2 | - | 4 - 5 | 771 | 97 | ||
| JN255799.1 | - | 4 - 5 | 506 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Calr4
Genomic Map