Assay Details
| Gene Symbol: Rab3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023537.5 | NP_076026.1 | 1 - 2 | 148 | 67 | |
| GenBank mRNA | AK012165.1 | - | 1 - 2 | 108 | 67 | |
| AK082959.1 | - | 1 - 2 | 148 | 67 | ||
| BC051918.1 | - | 1 - 2 | 53 | 5691723 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK012165.1, AK082959.1, BC051918.1, NM_023537.5 transcripts.
Set Membership: |
|

Rab3b
Genomic Map