Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400008420.mm.1 |
NM_001159964.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008420.mm.1 |
NM_001159964.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008420.mm.1 |
NM_007943.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008420.mm.1 |
NM_007943.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008420.mm.1 |
TC0400001164.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008417.mm.1 |
NM_001159964.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008417.mm.1 |
NM_001159964.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008417.mm.1 |
TC0400001164.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008418.mm.1 |
NM_001159964.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008418.mm.1 |
NM_001159964.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008418.mm.1 |
TC0400001164.mm.1 |
MTA TC |