Assay Details
| Gene Symbol: Afap1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178928.4 | NP_849259.2 | 2 - 3 | 218 | 97 | |
| GenBank mRNA | AK035227.1 | - | 2 - 3 | 218 | 97 | |
| AK053714.1 | - | 2 - 3 | 217 | 97 | ||
| BC094505.1 | - | 2 - 3 | 167 | 97 | ||
| BC138923.1 | - | 2 - 3 | 179 | 8861945 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Afap1l1
Genomic Map