Assay Details
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 12 - 13 | 1628 | 81 | |
| GenBank mRNA | AK031548.1 | - | 12 - 13 | 1628 | 81 | |
| AK040377.1 | - | 11 - 12 | 1859 | 81 | ||
| AK166470.1 | - | 10 - 11 | 1627 | 81 | ||
| BC023786.1 | - | 11 - 12 | 1006 | 81 | ||
| BC026832.1 | - | 12 - 13 | 1678 | 4486176 | 81 | |
| BC029977.1 | - | 3 - 4 | 264 | 81 | ||
| BC036128.1 | - | 12 - 13 | 1746 | 4971791 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrd1
Genomic Map