Assay Details
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 15 - 16 | 1835 | 75 | |
| GenBank mRNA | AK031548.1 | - | 15 - 16 | 1835 | 75 | |
| AK040377.1 | - | 14 - 15 | 2066 | 75 | ||
| AK050944.1 | - | 2 - 3 | 138 | 75 | ||
| AK166470.1 | - | 13 - 14 | 1834 | 75 | ||
| BC023786.1 | - | 14 - 15 | 1213 | 75 | ||
| BC026832.1 | - | 15 - 16 | 1885 | 4486176 | 75 | |
| BC029977.1 | - | 6 - 7 | 471 | 75 | ||
| BC036128.1 | - | 15 - 16 | 1953 | 4971791 | 75 | |
| U86112.1 | - | 1 - 2 | 83 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nrd1
Genomic Map