Assay Details
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 17 - 18 | 2056 | 151 | |
| GenBank mRNA | AK031548.1 | - | 17 - 18 | 2056 | 151 | |
| AK166470.1 | - | 15 - 16 | 2055 | 151 | ||
| BC023786.1 | - | 16 - 17 | 1434 | 151 | ||
| BC026832.1 | - | 17 - 18 | 2106 | 4486176 | 151 | |
| BC029977.1 | - | 8 - 9 | 692 | 151 | ||
| BC036128.1 | - | 17 - 18 | 2174 | 4971791 | 151 | |
| U86112.1 | - | 3 - 4 | 304 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nrd1
Genomic Map