Assay Details
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 18 - 19 | 2172 | 105 | |
| GenBank mRNA | AK031548.1 | - | 18 - 19 | 2172 | 105 | |
| AK166470.1 | - | 16 - 17 | 2171 | 105 | ||
| BC023786.1 | - | 17 - 18 | 1550 | 105 | ||
| BC024544.1 | - | 2 - 3 | 157 | 105 | ||
| BC026832.1 | - | 18 - 19 | 2222 | 4486176 | 105 | |
| BC029977.1 | - | 9 - 10 | 808 | 105 | ||
| BC036128.1 | - | 18 - 19 | 2290 | 4971791 | 105 | |
| U86112.1 | - | 4 - 5 | 420 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nrd1
Genomic Map