Assay Details
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 22 - 23 | 2663 | 74 | |
| GenBank mRNA | AK031548.1 | - | 22 - 23 | 2663 | 74 | |
| AK050944.1 | - | 8 - 9 | 846 | 74 | ||
| AK166470.1 | - | 20 - 21 | 2662 | 74 | ||
| BC023786.1 | - | 21 - 22 | 2041 | 74 | ||
| BC024544.1 | - | 6 - 7 | 648 | 74 | ||
| BC026832.1 | - | 22 - 23 | 2713 | 4486176 | 74 | |
| BC029977.1 | - | 13 - 14 | 1299 | 74 | ||
| BC036128.1 | - | 22 - 23 | 2781 | 4971791 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrd1
Genomic Map