Assay Details
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 23 - 24 | 2694 | 88 | |
| GenBank mRNA | AK031548.1 | - | 23 - 24 | 2694 | 88 | |
| AK050944.1 | - | 9 - 10 | 877 | 88 | ||
| AK166470.1 | - | 21 - 22 | 2693 | 88 | ||
| BC023786.1 | - | 22 - 23 | 2072 | 88 | ||
| BC024544.1 | - | 7 - 8 | 679 | 88 | ||
| BC026832.1 | - | 23 - 24 | 2744 | 4486176 | 88 | |
| BC029977.1 | - | 14 - 15 | 1330 | 88 | ||
| BC036128.1 | - | 23 - 24 | 2812 | 4971791 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrd1
Genomic Map