Assay Details
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 26 - 27 | 2937 | 99 | |
| GenBank mRNA | AK031548.1 | - | 26 - 27 | 2937 | 99 | |
| AK166470.1 | - | 24 - 25 | 2936 | 99 | ||
| BC023786.1 | - | 25 - 26 | 2315 | 99 | ||
| BC024544.1 | - | 10 - 11 | 922 | 99 | ||
| BC026832.1 | - | 26 - 27 | 2987 | 4486176 | 99 | |
| BC029977.1 | - | 17 - 18 | 1573 | 99 | ||
| BC036128.1 | - | 26 - 27 | 3055 | 4971791 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrd1
Genomic Map