Assay Details
| Gene Symbol: Slc5a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145551.4 | NP_663526.3 | 10 - 11 | 1267 | 73 | |
| GenBank mRNA | AK029158.1 | - | 10 - 11 | 1268 | 73 | |
| AK033425.1 | - | 9 - 10 | 1197 | 73 | ||
| AK050696.1 | - | 9 - 10 | 1147 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a9
Genomic Map