Assay Details
| Gene Symbol: Slc5a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145551.4 | NP_663526.3 | 7 - 8 | 822 | 85 | |
| GenBank mRNA | AK029158.1 | - | 7 - 8 | 823 | 85 | |
| AK033425.1 | - | 6 - 7 | 752 | 85 | ||
| AK050696.1 | - | 6 - 7 | 702 | 85 | ||
| BC021357.1 | - | 7 - 8 | 803 | 5012356 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a9
Genomic Map