Assay Details
| Gene Symbol: Osbpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134791.2 | NP_001128263.1 | 16 - 17 | 1609 | 106 | |
| NM_133885.4 | NP_598646.4 | 19 - 20 | 1842 | 106 | ||
| NM_173350.3 | NP_775485.2 | 17 - 18 | 1648 | 106 | ||
| GenBank mRNA | AK131984.1 | - | 19 - 20 | 1814 | 107 | |
| AK172010.1 | - | 17 - 18 | 1635 | 106 | ||
| AK178534.1 | - | 2 - 3 | 158 | 106 | ||
| AK202959.1 | - | 2 - 3 | 158 | 106 | ||
| BC021507.1 | - | 15 - 16 | 1470 | 5347121 | 106 | |
| BC023759.1 | - | 19 - 20 | 1797 | 106 | ||
| BC026927.1 | - | 17 - 18 | 1531 | 5134780 | 106 | |
| BC089163.1 | - | 16 - 17 | 1550 | 5367603 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osbpl9
Genomic Map