Assay Details
| Gene Symbol: Slc2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019741.3 | NP_062715.2 | 11 - 12 | 1754 | 94 | |
| GenBank mRNA | AF161071.1 | - | 9 - 10 | 1115 | 94 | |
| AK029720.1 | - | 11 - 12 | 1755 | 94 | ||
| AK203162.1 | - | 2 - 3 | 166 | 94 | ||
| BC023500.1 | - | 9 - 10 | 1130 | 5043284 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc2a5
Genomic Map