Assay Details
| Gene Symbol: Slc2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019741.3 | NP_062715.2 | 12 - 13 | 1825 | 73 | |
| GenBank mRNA | AF161071.1 | - | 10 - 11 | 1186 | 73 | |
| AK029720.1 | - | 12 - 13 | 1826 | 73 | ||
| BC023500.1 | - | 10 - 11 | 1201 | 5043284 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc2a5
Genomic Map