Assay Details
| Gene Symbol: Slc2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019741.3 | NP_062715.2 | 5 - 6 | 939 | 76 | |
| GenBank mRNA | AK029720.1 | - | 5 - 6 | 940 | 76 | |
| BC023500.1 | - | 3 - 4 | 315 | 5043284 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc2a5
Genomic Map