Assay Details
| Gene Symbol: Slc2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019741.3 | NP_062715.2 | 9 - 10 | 1533 | 93 | |
| GenBank mRNA | AF161071.1 | - | 7 - 8 | 894 | 93 | |
| AK029720.1 | - | 9 - 10 | 1534 | 93 | ||
| BC023500.1 | - | 7 - 8 | 909 | 5043284 | 93 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc2a5
Genomic Map