Assay Details
| Gene Symbol: Rere | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085492.1 | NP_001078961.1 | 8 - 9 | 1185 | 91 | |
| GenBank mRNA | BC145182.1 | - | 7 - 8 | 955 | 91 | |
More Information
Set Membership: |
|

Rere
Genomic Map