Assay Details
| Gene Symbol: Zfp532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC094671.1 | - | 3 - 4 | 2743 | 30618961 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp532| Gene Symbol: Zfp532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC094671.1 | - | 3 - 4 | 2743 | 30618961 | 72 |
Set Membership: |
|