Assay Details
| Gene Symbol: Sec11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025468.2 | NP_079744.1 | 1 - 2 | 139 | 70 | |
| GenBank mRNA | AK003027.1 | - | 1 - 2 | 170 | 70 | |
| AK003411.1 | - | 1 - 2 | 151 | 70 | ||
| AK007641.1 | - | 1 - 2 | 140 | 70 | ||
| BC037187.1 | - | 1 - 2 | 182 | 3666130 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec11c
Genomic Map