Assay Details
| Gene Symbol: Ablim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164491.1 | NP_001157963.1 | 9 - 10 | 1079 | 60 | |
| NM_198649.3 | NP_941051.2 | 8 - 9 | 1398 | 60 | ||
| GenBank mRNA | AK086538.1 | - | 8 - 9 | 1178 | 60 | |
| AK173040.1 | - | 8 - 9 | 956 | 60 | ||
| BC060275.1 | - | 9 - 10 | 1050 | 6822626 | 60 | |
| BC094229.1 | - | 8 - 9 | 1372 | 30534236 | 60 | |
| CF171696.1 | - | 3 - 4 | 174 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ablim3
Genomic Map