Assay Details
| Gene Symbol: Csnk1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146087.2 | NP_666199.1 | 6 - 7 | 845 | 138 | |
| GenBank mRNA | AK076070.1 | - | 6 - 7 | 1112 | 138 | |
| AK078398.1 | - | 6 - 7 | 1094 | 138 | ||
| AK141421.1 | - | 6 - 7 | 1100 | 138 | ||
| AK146873.1 | - | 6 - 7 | 1115 | 138 | ||
| AK147165.1 | - | 6 - 7 | 1090 | 138 | ||
| AK152387.1 | - | 6 - 7 | 1090 | 138 | ||
| AK152793.1 | - | 6 - 7 | 1110 | 138 | ||
| AK159939.1 | - | 3 - 4 | 314 | 138 | ||
| AK166318.1 | - | 6 - 7 | 1105 | 138 | ||
| AK166429.1 | - | 6 - 7 | 1087 | 138 | ||
| AK167550.1 | - | 6 - 7 | 1113 | 138 | ||
| AK180731.1 | - | 3 - 4 | 288 | 138 | ||
| AK188801.1 | - | 3 - 4 | 275 | 138 | ||
| AK219793.1 | - | 3 - 4 | 256 | 138 | ||
| BC019740.1 | - | 6 - 7 | 845 | 3487913 | 138 | |
| BC025439.1 | - | 6 - 7 | 792 | 5038433 | 138 | |
| BC048081.1 | - | 6 - 7 | 878 | 6309504 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Csnk1a1
Genomic Map