Assay Details
| Gene Symbol: Casz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159344.1 | NP_001152816.1 | 12 - 13 | 3171 | 78 | |
| NM_027195.2 | NP_081471.2 | 12 - 13 | 3171 | 78 | ||
| GenBank mRNA | AK173341.1 | - | 7 - 8 | 1961 | 78 | |
| BC075673.1 | - | 12 - 13 | 3173 | 78 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Casz1
Genomic Map