Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400015913.mm.1 |
NM_001159344.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015913.mm.1 |
NM_001159344.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015913.mm.1 |
NM_027195.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015913.mm.1 |
NM_027195.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015913.mm.1 |
TC0400001938.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400015903.mm.1 |
NM_001159344.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015903.mm.1 |
NM_001159344.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015903.mm.1 |
NM_027195.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015903.mm.1 |
NM_027195.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015903.mm.1 |
TC0400001938.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400015912.mm.1 |
NM_001159344.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015912.mm.1 |
NM_001159344.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015912.mm.1 |
NM_027195.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015912.mm.1 |
NM_027195.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015912.mm.1 |
TC0400001938.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400015899.mm.1 |
NM_001159344.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015899.mm.1 |
NM_001159344.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015899.mm.1 |
NM_027195.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015899.mm.1 |
NM_027195.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015899.mm.1 |
TC0400001938.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400015900.mm.1 |
NM_001159344.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015900.mm.1 |
NM_001159344.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015900.mm.1 |
NM_027195.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015900.mm.1 |
NM_027195.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015900.mm.1 |
TC0400001938.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400015902.mm.1 |
NM_001159344.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015902.mm.1 |
NM_001159344.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015902.mm.1 |
NM_027195.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015902.mm.1 |
NM_027195.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015902.mm.1 |
TC0400001938.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400015906.mm.1 |
NM_001159344.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015906.mm.1 |
NM_001159344.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015906.mm.1 |
NM_027195.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015906.mm.1 |
NM_027195.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015906.mm.1 |
TC0400001938.mm.1 |
MTA TC |