Assay Details
| Gene Symbol: Cdk11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007661.3 | NP_031687.2 | 18 - 19 | 2154 | 86 | |
| GenBank mRNA | AK077668.1 | - | 18 - 19 | 2155 | 86 | |
| AK147133.1 | - | 18 - 19 | 2132 | 86 | ||
| BC025058.1 | - | 7 - 8 | 804 | 86 | ||
| BC052920.1 | - | 18 - 19 | 2232 | 6489832 | 86 | |
| L37092.1 | - | 17 - 18 | 2726 | 86 | ||
| M58633.1 | - | 13 - 14 | 1497 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk11b
Genomic Map