Assay Details
| Gene Symbol: Cdk11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007661.3 | NP_031687.2 | 4 - 5 | 447 | 69 | |
| GenBank mRNA | AK077668.1 | - | 4 - 5 | 448 | 69 | |
| AK135402.1 | - | 4 - 5 | 454 | 69 | ||
| AK147133.1 | - | 4 - 5 | 425 | 69 | ||
| BC052920.1 | - | 4 - 5 | 525 | 6489832 | 69 | |
| L37092.1 | - | 3 - 4 | 1022 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk11b
Genomic Map