| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100021251.mm.1 |
NM_001110831.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100021251.mm.1 |
NM_001110831.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100021251.mm.1 |
NM_016878.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100021251.mm.1 |
NM_016878.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100021251.mm.1 |
TC0100002608.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100021252.mm.1 |
NM_001110831.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100021252.mm.1 |
NM_001110831.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100021252.mm.1 |
NM_016878.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100021252.mm.1 |
NM_016878.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100021252.mm.1 |
TC0100002608.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100021253.mm.1 |
NM_001110831.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100021253.mm.1 |
NM_001110831.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100021253.mm.1 |
NM_016878.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100021253.mm.1 |
NM_016878.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100021253.mm.1 |
TC0100002608.mm.1 |
MTA TC |