Assay Details
| Gene Symbol: Dnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110831.1 | NP_001104301.1 | 7 - 8 | 932 | 96 | |
| NM_016878.4 | NP_058574.3 | 7 - 8 | 756 | 96 | ||
| GenBank mRNA | AF005051.1 | - | 6 - 7 | 866 | 96 | |
| AK038646.1 | - | 932 | 96 | |||
| AK052001.1 | - | 734 | 96 | |||
| AK160084.1 | - | 7 - 8 | 693 | 96 | ||
| AK166276.1 | - | 7 - 8 | 706 | 96 | ||
| BC092232.1 | - | 7 - 8 | 684 | 30551654 | 96 | |
More Information
Set Membership: |
|

Dnpep
Genomic Map