Assay Details
| Gene Symbol: Dnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110831.1 | NP_001104301.1 | 8 - 9 | 1037 | 95 | |
| NM_016878.4 | NP_058574.3 | 8 - 9 | 861 | 95 | ||
| GenBank mRNA | AF005051.1 | - | 7 - 8 | 971 | 95 | |
| AK052001.1 | - | 839 | 95 | |||
| AK160084.1 | - | 8 - 9 | 798 | 95 | ||
| AK166276.1 | - | 8 - 9 | 811 | 95 | ||
| AK215853.1 | - | 1 - 2 | 98 | 95 | ||
| BC092232.1 | - | 8 - 9 | 789 | 30551654 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnpep
Genomic Map