Assay Details
| Gene Symbol: Dnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110831.1 | NP_001104301.1 | 10 - 11 | 1207 | 64 | |
| NM_016878.4 | NP_058574.3 | 10 - 11 | 1031 | 64 | ||
| GenBank mRNA | AF005051.1 | - | 9 - 10 | 1141 | 64 | |
| AK038646.1 | - | 1207 | 64 | |||
| AK052001.1 | - | 1009 | 64 | |||
| AK160084.1 | - | 10 - 11 | 968 | 64 | ||
| AK166276.1 | - | 10 - 11 | 981 | 64 | ||
| BC092232.1 | - | 10 - 11 | 959 | 30551654 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnpep
Genomic Map