Assay Details
| Gene Symbol: Tuba4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009447.3 | NP_033473.1 | 1 - 2 | 61 | 68 | |
| GenBank mRNA | AK002427.1 | - | 1 - 2 | 62 | 68 | |
| AK078355.1 | - | 1 - 2 | 62 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK002427.1,AK078355.1,NM_009447.3 transcripts.
This assay is in the 5'UTR region of the AK002427.1,AK078355.1,NM_009447.3 transcripts.
Set Membership: |
|

Tuba4a
Genomic Map