Assay Details
| Gene Symbol: Ptprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008985.2 | NP_033011.2 | 1 - 2 | 202 | 87 | |
| GenBank mRNA | AK041296.1 | - | 1 - 2 | 208 | 87 | |
| AK076223.1 | - | 1 - 2 | 181 | 87 | ||
| AK139398.1 | - | 1 - 2 | 202 | 87 | ||
| AK163141.1 | - | 1 - 2 | 178 | 87 | ||
| AK220467.1 | - | 1 - 2 | 160 | 87 | ||
| BB869850.1 | - | 1 - 2 | 146 | 87 | ||
| BC068165.1 | - | 1 - 2 | 134 | 87 | ||
| DQ832283.1 | - | 1 - 2 | 179 | 87 | ||
| U11812.1 | - | 1 - 2 | 126 | 87 | ||
| X74438.1 | - | 1 - 2 | 219 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK139398.1 transcripts.
Set Membership: |
|

Ptprn
Genomic Map