Assay Details
| Gene Symbol: Plcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081456.1 | NP_001074925.1 | 11 - 12 | 1821 | 62 | |
| NM_148937.2 | NP_683739.2 | 12 - 13 | 2068 | 62 | ||
| GenBank mRNA | AK029561.1 | - | 12 - 13 | 2097 | 62 | |
| AK140231.1 | - | 11 - 12 | 1805 | 62 | ||
| AY033991.1 | - | 12 - 13 | 1900 | 62 | ||
| BC066156.1 | - | 11 - 12 | 1712 | 30099811 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plcd4
Genomic Map