Assay Details
| Gene Symbol: Plcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081456.1 | NP_001074925.1 | 2 - 3 | 195 | 81 | |
| NM_148937.2 | NP_683739.2 | 2 - 3 | 346 | 81 | ||
| GenBank mRNA | AK016945.1 | - | 2 - 3 | 343 | 81 | |
| AK029561.1 | - | 2 - 3 | 341 | 81 | ||
| AK039149.1 | - | 2 - 3 | 173 | 81 | ||
| AK140231.1 | - | 2 - 3 | 182 | 81 | ||
| AY033991.1 | - | 2 - 3 | 178 | 81 | ||
| BC066156.1 | - | 2 - 3 | 86 | 30099811 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016945.1, AK039149.1, AK140231.1, AY033991.1, BC066156.1, NM_001081456.1, NM_148937.2 transcripts.
Set Membership: |
|

Plcd4
Genomic Map