Assay Details
| Gene Symbol: Zfand2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159905.1 | NP_001153377.1 | 3 - 4 | 228 | 63 | |
| NM_001159906.1 | NP_001153378.1 | 3 - 4 | 225 | 63 | ||
| NM_026846.3 | NP_081122.2 | 2 - 3 | 262 | 63 | ||
| GenBank mRNA | AK004332.1 | - | 2 - 3 | 252 | 63 | |
| AK151430.1 | - | 2 - 3 | 264 | 63 | ||
| BC011495.1 | - | 2 - 3 | 287 | 4196340 | 63 | |
| BY197482.1 | - | 3 - 4 | 226 | 63 | ||
| CX217579.1 | - | 3 - 4 | 231 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfand2b
Genomic Map