Assay Details
| Gene Symbol: Stk11ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027886.3 | NP_082162.3 | 11 - 12 | 1126 | 78 | |
| GenBank mRNA | AK004757.2 | - | 11 - 12 | 1079 | 78 | |
| AK156122.1 | - | 10 - 11 | 1214 | 78 | ||
| AK165911.1 | - | 11 - 12 | 1117 | 78 | ||
| AK171989.1 | - | 10 - 11 | 988 | 78 | ||
| BC042403.1 | - | 9 - 10 | 1163 | 78 | ||
| BC061635.1 | - | 11 - 12 | 1109 | 6530178 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Stk11ip
Genomic Map