Assay Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170755.2 | NP_739561.2 | 4 - 5 | 616 | 93 | |
| GenBank mRNA | AK034667.1 | - | 4 - 5 | 617 | 93 | |
| AK152268.1 | - | 4 - 5 | 331 | 93 | ||
| AK158940.1 | - | 4 - 5 | 386 | 93 | ||
| AK160400.1 | - | 4 - 5 | 570 | 93 | ||
| AK165957.1 | - | 4 - 5 | 595 | 93 | ||
| AK172093.1 | - | 4 - 5 | 613 | 93 | ||
| BC070406.1 | - | 4 - 5 | 608 | 5707319 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam134a
Genomic Map