Assay Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170755.2 | NP_739561.2 | 7 - 8 | 942 | 56 | |
| GenBank mRNA | AK034667.1 | - | 7 - 8 | 944 | 56 | |
| AK152268.1 | - | 7 - 8 | 657 | 56 | ||
| AK160400.1 | - | 7 - 8 | 896 | 56 | ||
| AK165957.1 | - | 7 - 8 | 921 | 56 | ||
| AK172093.1 | - | 7 - 8 | 939 | 56 | ||
| AK210767.1 | - | 1 - 2 | 51 | 56 | ||
| BC026519.1 | - | 3 - 4 | 293 | 56 | ||
| BC038286.1 | - | 3 - 4 | 328 | 4221441 | 56 | |
| BC070406.1 | - | 7 - 8 | 934 | 5707319 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam134a
Genomic Map