Assay Details
| Gene Symbol: Glb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029010.1 | NP_083286.1 | 14 - 15 | 1581 | 68 | |
| GenBank mRNA | AK014667.1 | - | 12 - 13 | 1604 | 68 | |
| BC021773.1 | - | 13 - 14 | 1334 | 4222994 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glb1l
Genomic Map