Assay Details
| Gene Symbol: Ttll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014974.1 | NP_001014974.1 | 16 - 17 | 3368 | 100 | |
| GenBank mRNA | AK129075.1 | - | 14 - 15 | 2195 | 100 | |
| AM690747.1 | - | 14 - 15 | 2971 | 100 | ||
| BC044790.1 | - | 3 - 4 | 345 | 100 | ||
| BC049820.1 | - | 1 - 2 | 599 | 100 | ||
| BC085151.1 | - | 16 - 17 | 3368 | 30534216 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttll4
Genomic Map