Assay Details
| Gene Symbol: Ttll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014974.1 | NP_001014974.1 | 17 - 18 | 3580 | 70 | |
| GenBank mRNA | AK129075.1 | - | 15 - 16 | 2407 | 70 | |
| AM690747.1 | - | 15 - 16 | 3183 | 70 | ||
| BC044790.1 | - | 4 - 5 | 557 | 70 | ||
| BC049820.1 | - | 2 - 3 | 811 | 70 | ||
| BC085151.1 | - | 17 - 18 | 3580 | 30534216 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttll4
Genomic Map